Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023671.2 | NP_076160.1 | 7 | 892 | ||
GenBank mRNA | AK011691.1 | - | 7 | 872 | ||
AK011789.1 | - | 7 | 877 | |||
AK031877.1 | - | 7 | 874 | |||
AK036156.1 | - | 1 | 387 | |||
AK078050.1 | - | 7 | 845 | |||
AK133981.1 | - | 7 | 885 | |||
AK151356.1 | - | 7 | 877 | |||
AK151944.1 | - | 7 | 877 | |||
AK169259.1 | - | 7 | 856 | |||
AK185541.1 | - | 1 | 46 | |||
BC005575.1 | - | 7 | 858 | 3967933 |