Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023709.4 | NP_076198.2 | 14 | 1662 | ||
GenBank mRNA | AK008629.1 | - | 14 | 1663 | ||
AK144514.1 | - | 14 | 1665 | |||
BC058748.1 | - | 14 | 1640 | 6394969 | ||
U89513.1 | - | 14 | 1623 |