Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023732.2 | NP_076221.1 | 10, 11 | 1828 | ||
GenBank mRNA | AK004605.1 | - | 10, 11 | 1908 | ||
AK040323.1 | - | 9 | 2065 | |||
AK151165.1 | - | 10, 11 | 1937 | |||
AK164003.1 | - | 10, 11 | 1832 | |||
AK168642.1 | - | 10, 11 | 1929 | |||
BC006634.1 | - | 10, 11 | 1828 | 3710552 |