Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205385.1 | NP_001192314.1 | 13 | 1559 | ||
NM_001205386.1 | NP_001192315.1 | 13 | 1556 | |||
NM_023735.2 | NP_076224.1 | 12 | 1751 | |||
GenBank mRNA | AK004554.1 | - | 12 | 1565 | ||
AK083343.1 | - | 12 | 1569 | |||
AK145536.1 | - | 12 | 1571 | |||
AK150487.1 | - | 12 | 1453 | |||
AK150589.1 | - | 12 | 1529 | |||
AK150991.1 | - | 12 | 1508 | |||
AK152771.1 | - | 1 | 114 | |||
AK152865.1 | - | 12 | 1545 | |||
AK153079.1 | - | 12 | 1572 | |||
AK153150.1 | - | 12 | 1546 | |||
AK153204.1 | - | 12 | 1452 | |||
AK159135.1 | - | 12 | 1538 | |||
AK159668.1 | - | 12 | 1463 | |||
AK159775.1 | - | 12 | 1550 | |||
AK160047.1 | - | 12 | 1411 | |||
AK166529.1 | - | 13 | 1503 | |||
AK166586.1 | - | 13 | 1503 | |||
AK167148.1 | - | 12 | 1510 | |||
AK167891.1 | - | 12 | 1508 | |||
AK168568.1 | - | 12 | 1438 | |||
AK168771.1 | - | 12 | 1524 | |||
AK177697.1 | - | 2 | 233 | |||
BC005557.1 | - | 12 | 1514 | 3156748 | ||
BC080806.1 | - | 13 | 1517 | 5350752 | ||
BC108386.1 | - | 12 | 1482 | 5051125 |