Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290448.1 | NP_001277377.1 | 2 | 457 | ||
NM_001290449.1 | NP_001277378.1 | 2 | 457 | |||
NM_023739.3 | NP_076228.2 | 2 | 420 | |||
GenBank mRNA | AF223576.1 | - | 2 | 384 | ||
AK005038.2 | - | 2 | 392 | |||
AK033850.1 | - | 2 | 351 | |||
AK050749.1 | - | 2 | 393 | |||
AK144647.1 | - | 2 | 406 | |||
AK146108.1 | - | 2 | 374 | |||
AK155387.1 | - | 2 | 372 | |||
AK160013.1 | - | 2 | 390 | |||
BC053709.1 | - | 2 | 355 | 30052746 | ||
CK388342.1 | - | 2 | 457 |