Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023740.2 | NP_076229.2 | 11 | 1614 | ||
GenBank mRNA | AF176814.1 | - | 11 | 1587 | ||
AK005255.1 | - | 10 | 1346 | |||
AK151505.1 | - | 11 | 1596 | |||
AK168042.1 | - | 11 | 1589 | |||
AK178032.1 | - | 1 | 252 | |||
AK179256.1 | - | 1 | 261 | |||
AK185803.1 | - | 1 | 286 | |||
AK196943.1 | - | 1 | 285 | |||
AK200562.1 | - | 1 | 221 | |||
AK214809.1 | - | 1 | 285 | |||
AK219851.1 | - | 1 | 285 | |||
BC010835.1 | - | 11 | 1578 | 4239913 |