Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166547.1 | NP_001160019.1 | 7 | 1140 | ||
NM_001166548.1 | NP_001160020.1 | 7 | 1144 | |||
NM_001166549.1 | NP_001160021.1 | 7 | 1248 | |||
NM_023743.2 | NP_076232.2 | 7 | 1155 | |||
GenBank mRNA | AB031388.1 | - | 7 | 976 | ||
AK041598.1 | - | 7 | 1106 | |||
AK088425.1 | - | 2 | 242 | |||
AK128990.1 | - | 7 | 1246 | |||
AK162367.1 | - | 1 | 87 | |||
AK165678.1 | - | 7 | 1143 | |||
AK205705.1 | - | 2 | 321 | |||
BC033410.1 | - | 7 | 990 | 5359290 |