Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023764.3 | NP_076253.1 | 6 | 2865 | ||
GenBank mRNA | AK016482.1 | - | 6 | 2866 | ||
AK049198.1 | - | 1 | 879 | |||
AK161312.1 | - | 6 | 2852 | |||
BC053515.1 | - | 6 | 2852 | |||
BC062139.1 | - | 6 | 2846 | 6334946 |