Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023794.2 | NP_076283.2 | 13 | 3347 | ||
GenBank mRNA | AK003461.1 | - | 1 | 842 | ||
AK049234.1 | - | 13 | 3397 | |||
AK080674.1 | - | 13 | 3324 | |||
AK164091.1 | - | 13 | 3397 | |||
AK168881.1 | - | 13 | 3394 | |||
BC034680.1 | - | 13 | 3347 | 4036564 | ||
BC039044.1 | - | 1 | 159 |