Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023799.3 | NP_076288.1 | 4 | 587 | ||
GenBank mRNA | AF132214.1 | - | 4 | 432 | ||
AK088774.1 | - | 4 | 850 | |||
AK129188.1 | - | 4 | 816 | |||
AK145227.1 | - | 4 | 837 | |||
BC020093.1 | - | 4 | 524 | |||
BC041109.1 | - | 4 | 917 | |||
BC054821.1 | - | 4 | 587 | 6515250 |