Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025438.2 | NP_001020609.1 | 15, 16 | 1598 | ||
NM_001025439.2 | NP_001020610.1 | 17, 18 | 1700 | |||
NM_001293663.1 | NP_001280592.1 | 17, 18 | 1700 | |||
NM_001293664.1 | NP_001280593.1 | 16, 17 | 1640 | |||
NM_001293665.1 | NP_001280594.1 | 16, 17 | 1591 | |||
NM_023813.4 | NP_076302.1 | 15, 16 | 1598 | |||
GenBank mRNA | AF059029.1 | - | 2, 3 | 133 | ||
AK012702.1 | - | 15, 16 | 1219 | |||
AK039076.1 | - | 17, 18 | 1309 | |||
AK044264.1 | - | 9, 10 | 626 | |||
AK132185.1 | - | 2, 3 | 117 | |||
AK142435.1 | - | 15, 16 | 1541 | |||
AK145106.1 | - | 17, 18 | 1629 | |||
AK148840.1 | - | 16, 17 | 1347 | |||
AK209911.1 | - | 1, 2 | 84 | |||
AK220516.1 | - | 17, 18 | 1700 | |||
BC042895.1 | - | 15, 16 | 2139 | |||
BC052894.1 | - | 15, 16 | 1209 | 30055337 | ||
BC057103.1 | - | 16, 17 | 1507 |