Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023815.4 | NP_076304.2 | 2 | 970 | ||
GenBank mRNA | AB028045.1 | - | 2 | 960 | ||
AK011574.2 | - | 2 | 931 | |||
AK028411.1 | - | 2 | 971 | |||
AK046261.1 | - | 2 | 948 | |||
AK049291.1 | - | 2 | 955 | |||
AK160279.1 | - | 2 | 950 | |||
BC017155.1 | - | 2 | 988 | 4910610 |