Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159965.1 | NP_001153437.1 | 3 | 319 | ||
NM_001159966.1 | NP_001153438.1 | 3 | 217 | |||
NM_001159967.1 | NP_001153439.1 | 3 | 319 | |||
NM_023884.4 | NP_076373.3 | 3 | 259 | |||
GenBank mRNA | AF312924.1 | - | 2 | 167 | ||
AK018622.1 | - | 3 | 261 | |||
AK029049.1 | - | 3 | 260 | |||
AK029993.1 | - | 3 | 292 | |||
AK033549.1 | - | 3 | 215 | |||
AK161080.1 | - | 3 | 286 | |||
AK165002.1 | - | 3 | 211 | |||
AK169368.1 | - | 3 | 237 | |||
BC043132.1 | - | 3 | 224 | 6811441 | ||
BC052663.1 | - | 3 | 236 | 30051209 | ||
CJ104104.1 | - | 3 | 319 |