Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023906.3 | NP_076395.2 | 2 | 273 | ||
GenBank mRNA | AF155354.1 | - | 1 | 88 | ||
AK010315.1 | - | 2 | 209 | |||
AK028489.1 | - | 2 | 266 | |||
AK038086.1 | - | 2 | 254 | |||
AK038897.1 | - | 2 | 224 | |||
AK143580.1 | - | 2 | 208 | |||
BB618690.1 | - | 2 | 275 | |||
BC023086.1 | - | 2 | 207 | 4947610 |