Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138392.3 | NP_612401.2 | 18 | 2274 | ||
XM_006723474.1 | XP_006723537.1 | 18 | 2304 | |||
XM_006723475.1 | XP_006723538.1 | 18 | 2301 | |||
XM_011527492.1 | XP_011525794.1 | 15 | 1953 | |||
XM_011527493.1 | XP_011525795.1 | 17 | 2200 | |||
XM_017027473.1 | XP_016882962.1 | 18 | 2271 | |||
XM_017027474.1 | XP_016882963.1 | 17 | 2170 | |||
GenBank mRNA | AF258553.1 | - | 18 | 2273 | ||
AK075057.1 | - | 18 | 2196 | |||
AK123695.1 | - | 16 | 2252 | |||
AK297006.1 | - | 12 | 1900 | |||
AK300808.1 | - | 14 | 1697 | |||
AY007117.1 | - | 9 | 1309 | |||
BC007653.1 | - | 10 | 1560 | |||
BC022855.1 | - | 18 | 2245 | 4102901 |