Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 1, 2 | 310 | ||
GenBank mRNA | AK002900.1 | - | 1, 2 | 261 | ||
AK009891.1 | - | 1, 2 | 289 | |||
AK010050.1 | - | 1, 2 | 286 | |||
AK049694.1 | - | 1, 2 | 292 | |||
AK049869.1 | - | 1, 2 | 286 | |||
AK075792.1 | - | 1, 2 | 303 | |||
AK143250.1 | - | 1, 2 | 282 | |||
BC002225.1 | - | 1, 2 | 294 | |||
BC027202.1 | - | 1, 2 | 214 | 4217391 | ||
BY297479.1 | - | 1, 2 | 321 |