Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024189.6 | NP_077151.3 | 2 | 266 | ||
GenBank mRNA | AB100452.1 | - | 2 | 256 | ||
AB100453.1 | - | 2 | 256 | |||
AK005008.1 | - | 2 | 250 | |||
AK011702.1 | - | 2 | 267 | |||
AK012787.1 | - | 1 | 102 | |||
AK148634.1 | - | 2 | 210 | |||
BC002192.1 | - | 2 | 139 | 3488691 |