Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cuedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164290.1 | NP_001157762.1 | 4 | 354 | ||
NM_001164291.1 | NP_001157763.1 | 3 | 246 | |||
NM_001164292.1 | NP_001157764.1 | 3 | 243 | |||
NM_001164293.1 | NP_001157765.1 | 3 | 246 | |||
NM_001164294.1 | NP_001157766.1 | 3 | 246 | |||
NM_001164295.1 | NP_001157767.1 | 3 | 243 | |||
NM_024192.2 | NP_077154.2 | 4 | 345 | |||
GenBank mRNA | AB232359.1 | - | 2 | 144 | ||
AK008842.1 | - | 3 | 209 | |||
AK013884.1 | - | 3 | 204 | |||
AK186231.1 | - | 3 | 169 | |||
AK207375.1 | - | 4 | 271 | |||
BC003442.1 | - | 4 | 309 | 2650719 | ||
BY279028.1 | - | 3 | 245 | |||
BY312150.1 | - | 3 | 207 | |||
CK625575.1 | - | 3 | 172 | |||
CX207621.1 | - | 3 | 545 | |||
CX218143.1 | - | 4 | 284 | |||
CX223880.1 | - | 4 | 310 |