Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139126.3 | NP_624311.1 | 11, 12 | 1182 | ||
GenBank mRNA | AF068298.1 | - | 2, 3 | 145 | ||
AF308286.1 | - | 1, 2 | 70 | |||
AF357880.1 | - | 11, 12 | 1115 | |||
AK315388.1 | - | 11, 12 | 1182 | |||
BC016984.1 | - | 3, 4 | 275 | |||
BC020986.1 | - | 11, 12 | 1118 | 3852013 | ||
BC064134.1 | - | 11, 12 | 1100 | |||
BC107684.1 | - | 11, 12 | 1116 | |||
BX537536.1 | - | 11, 12 | 1117 |