Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024204.6 | NP_077166.4 | 2 | 456 | ||
GenBank mRNA | AK012869.1 | - | 2 | 469 | ||
AK017360.1 | - | 2 | 457 | |||
AK017408.1 | - | 2 | 341 | |||
AK037242.1 | - | 2 | 339 | |||
BC003460.1 | - | 2 | 318 | 2648395 |