Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024206.4 | NP_077168.2 | 4 | 303 | ||
GenBank mRNA | AK003354.1 | - | 4 | 304 | ||
AK009755.1 | - | 3 | 287 | |||
BC002128.1 | - | 4 | 306 | 3590767 |