Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024226.4 | NP_077188.1 | 2 | 489 | ||
NM_194051.3 | NP_918940.1 | 4 | 2776 | |||
NM_194052.3 | NP_918941.1 | 2 | 965 | |||
NM_194053.3 | NP_918942.1 | 3 | 1022 | |||
NM_194054.3 | NP_918943.1 | 4 | 3383 | |||
GenBank mRNA | AB073672.1 | - | 2 | 1341 | ||
AF326337.1 | - | 2 | 269 | |||
AK034902.1 | - | 2 | 909 | |||
AK211699.1 | - | 2 | 205 | |||
AK212390.1 | - | 2 | 192 | |||
AK220511.1 | - | 4 | 3343 | |||
AY102280.1 | - | 4 | 2776 | |||
AY102281.1 | - | 2 | 922 | |||
AY102282.1 | - | 3 | 979 | |||
AY102283.1 | - | 2 | 482 | |||
AY102284.1 | - | 4 | 3340 | |||
AY114152.1 | - | 4 | 3341 | |||
AY114153.1 | - | 3 | 796 | |||
BC032192.1 | - | 2 | 1635 | 5323152 | ||
BC032272.1 | - | 2 | 2489 | |||
BC056373.1 | - | 4 | 3211 | |||
BU848611.1 | - | 2 | 328 |