Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcyt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024229.2 | NP_077191.2 | 2, 3 | 226 | ||
GenBank mRNA | AK140458.1 | - | 2, 3 | 266 | ||
AK144379.1 | - | 2 | 602 | |||
AK144544.1 | - | 2, 3 | 273 | |||
AK166319.1 | - | 2, 3 | 255 | |||
BC003473.1 | - | 2, 3 | 232 | 2650077 | ||
BC008276.1 | - | 2, 3 | 226 | 3707732 |