Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170489.1 | NP_001163960.1 | 4 | 663 | ||
NM_024251.4 | NP_077213.2 | 4 | 533 | |||
GenBank mRNA | AK008513.1 | - | 4 | 664 | ||
AK034191.1 | - | 4 | 513 | |||
AK078805.1 | - | 4 | 626 | |||
BC002179.1 | - | 4 | 311 | 3487226 |