Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec61b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024171.2 | NP_077133.1 | 2, 3 | 199 | ||
GenBank mRNA | AK004505.1 | - | 2, 3 | 200 | ||
AK008468.2 | - | 642 | ||||
AK008739.1 | - | 2, 3 | 203 | |||
BC002089.1 | - | 2, 3 | 171 | 3487410 | ||
BC081445.1 | - | 2, 3 | 167 | 6412192 |