Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291080.1 | NP_001278009.1 | 5 | 676 | ||
NM_024176.2 | NP_077138.1 | 5 | 655 | |||
GenBank mRNA | AK005445.1 | - | 4 | 390 | ||
AK010152.1 | - | 5 | 609 | |||
AK087456.1 | - | 5 | 481 | |||
AK154251.1 | - | 4 | 435 | |||
BC002090.1 | - | 5 | 592 | 3488139 | ||
CT010290.1 | - | 5 | 390 |