Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024181.2 | NP_077143.2 | 3 | 820 | ||
GenBank mRNA | AF255459.1 | - | 3 | 497 | ||
AF314002.1 | - | 2 | 333 | |||
AK004617.1 | - | 3 | 575 | |||
AK013930.1 | - | 3 | 516 | |||
AK016269.1 | - | 3 | 820 | |||
AK028184.1 | - | 3 | 560 | |||
AK128945.1 | - | 3 | 575 | |||
AK165652.1 | - | 3 | 578 | |||
AK166502.1 | - | 3 | 530 | |||
AK169020.1 | - | 3 | 587 | |||
AK179556.1 | - | 1 | 69 | |||
BC033461.1 | - | 3 | 559 | 4010341 |