Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024182.4 | NP_077144.2 | 3 | 366 | ||
GenBank mRNA | AK004748.1 | - | 3 | 458 | ||
AK131635.1 | - | 3 | 370 | |||
AK134327.1 | - | 3 | 458 | |||
AK147074.1 | - | 3 | 367 | |||
AK167309.1 | - | 3 | 462 | |||
AK170882.1 | - | 3 | 446 | |||
BC033271.1 | - | 3 | 404 | 4016197 |