Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024206.4 | NP_077168.2 | 5 | 454 | ||
GenBank mRNA | AK003354.1 | - | 5 | 455 | ||
AK009755.1 | - | 4 | 438 | |||
BC002128.1 | - | 5 | 457 | 3590767 |
Gene Symbol: 4933417G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016844.1 | - | 1 | 259 |