Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024218.4 | NP_077180.1 | 3 | 337 | ||
GenBank mRNA | AK018731.1 | - | 3 | 220 | ||
AK019214.1 | - | 3 | 210 | |||
AK028209.1 | - | 3 | 228 | |||
AK077617.1 | - | 3 | 211 | |||
AK136631.1 | - | 3 | 220 | |||
BC002110.1 | - | 3 | 212 | |||
BC053377.1 | - | 3 | 208 | 4216355 | ||
BC058114.1 | - | 3 | 207 | 6817726 | ||
BC092008.1 | - | 3 | 203 | 30612436 | ||
BY031373.1 | - | 3 | 337 |