Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000019.3 | NP_000010.1 | 1, 2 | 156 | ||
XM_017017682.1 | XP_016873171.1 | 1, 2 | 120 | |||
GenBank mRNA | AK312574.1 | - | 1, 2 | 123 | ||
BC010942.1 | - | 1, 2 | 124 | |||
D90228.1 | - | 1, 2 | 156 |