Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038643.1 | NP_001033732.1 | 2 | 825 | ||
NM_023908.2 | NP_076397.2 | 2 | 825 | |||
GenBank mRNA | AF226324.1 | - | 2 | 723 | ||
AK037938.1 | - | 2 | 640 | |||
AK046945.1 | - | 1 | 185 | |||
AK151781.1 | - | 2 | 795 | |||
AK152488.1 | - | 2 | 659 | |||
BC025059.1 | - | 2 | 658 | 2598913 | ||
BC094464.1 | - | 2 | 617 |