Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ern1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023913.2 | NP_076402.1 | 11, 12 | 1337 | ||
GenBank mRNA | AB031332.1 | - | 11, 12 | 1337 | ||
AK077041.1 | - | 1358 | ||||
AK080709.1 | - | 5, 6 | 589 |