Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024124.3 | NP_077038.2 | 3, 4 | 603 | ||
GenBank mRNA | AF235053.1 | - | 2, 3 | 272 | ||
AF279371.1 | - | 2, 3 | 272 | |||
AF324492.1 | - | 2, 3 | 272 | |||
BC098187.1 | - | 3, 4 | 588 | 5363624 |