Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024177.3 | NP_077139.2 | 5 | 729 | ||
GenBank mRNA | AK088343.1 | - | 5 | 668 | ||
AK151598.1 | - | 5 | 668 | |||
AK159033.1 | - | 5 | 730 | |||
BC002319.1 | - | 4 | 705 | 3593984 | ||
BC030739.1 | - | 5 | 656 | 1493919 |