Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024180.5 | NP_077142.1 | 4 | 676 | ||
GenBank mRNA | AK002583.1 | - | 4 | 639 | ||
AK002824.1 | - | 3 | 383 | |||
AK007548.1 | - | 3 | 459 | |||
AK012681.1 | - | 3 | 459 | |||
AK013455.1 | - | 4 | 649 | |||
BC002146.1 | - | 4 | 697 | 3484833 | ||
DV065903.1 | - | 4 | 719 |