Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 9, 10 | 1014 | ||
GenBank mRNA | AK002900.1 | - | 9, 10 | 965 | ||
AK009891.1 | - | 9, 10 | 993 | |||
AK010050.1 | - | 9, 10 | 981 | |||
AK049869.1 | - | 8, 9 | 938 | |||
AK075792.1 | - | 7, 8 | 784 | |||
AK143250.1 | - | 9, 10 | 986 | |||
AK208803.1 | - | 3, 4 | 240 | |||
BC002225.1 | - | 9, 10 | 996 | |||
BC027202.1 | - | 9, 10 | 918 | 4217391 |