Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024216.1 | NP_077178.1 | 5, 6 | 674 | ||
GenBank mRNA | AK016996.1 | - | 5, 6 | 605 | ||
AK080720.1 | - | 5, 6 | 621 | |||
AK087968.1 | - | 5, 6 | 625 | |||
AK146783.1 | - | 5, 6 | 612 | |||
BC003341.1 | - | 5, 6 | 674 | 3257412 |