Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024237.4 | NP_077199.2 | 5, 6 | 821 | ||
GenBank mRNA | AK005465.1 | - | 5, 6 | 773 | ||
BC051550.1 | - | 3, 4 | 377 | |||
BC095941.1 | - | 5, 6 | 753 | 4167912 | ||
BM936626.1 | - | 1, 2 | 82 | |||
EF668007.1 | - | 5, 6 | 677 |