Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170489.1 | NP_001163960.1 | 6 | 942 | ||
NM_024251.4 | NP_077213.2 | 6 | 812 | |||
GenBank mRNA | AK008513.1 | - | 6 | 943 | ||
AK034191.1 | - | 6 | 792 | |||
AK078805.1 | - | 6 | 882 | |||
BC002179.1 | - | 6 | 590 | 3487226 |