Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024261.2 | NP_077223.2 | 2, 3 | 221 | ||
GenBank mRNA | AK006776.1 | - | 2, 3 | 231 | ||
AK134431.1 | - | 2, 3 | 188 | |||
AK209106.1 | - | 1, 2 | 79 | |||
BC002139.1 | - | 2, 3 | 206 | |||
BC099894.1 | - | 2, 3 | 314 | |||
BC138908.1 | - | 2, 3 | 181 | 8861930 | ||
BC145732.1 | - | 2, 3 | 181 | 40131035 | ||
BU148062.1 | - | 2, 3 | 206 |