Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000034.3 | NP_000025.1 | 13 | 2070 | ||
NM_001127617.2 | NP_001121089.1 | 8 | 1292 | |||
NM_001243177.1 | NP_001230106.1 | 9 | 1413 | |||
NM_184041.2 | NP_908930.1 | 8 | 1259 | |||
NM_184043.2 | NP_908932.1 | 8 | 1231 | |||
XM_011545768.2 | XP_011544070.1 | 10 | 1933 | |||
GenBank mRNA | AK026577.1 | - | 8 | 1108 | ||
AK098778.1 | - | 11 | 2111 | |||
AK301993.1 | - | 9 | 1225 | |||
BC000367.2 | - | 5 | 671 | |||
BC004333.1 | - | 13 | 1990 | 3630974 | ||
BC010660.1 | - | 8 | 1177 | 3869603 | ||
BC012880.1 | - | 8 | 1105 | 3459009 | ||
BC013614.1 | - | 8 | 1188 | 3880857 | ||
BC015888.2 | - | 8 | 1101 | 3911623 | ||
BC016170.1 | - | 5 | 671 | |||
BC016800.1 | - | 9 | 1129 | 4072487 | ||
BU855133.1 | - | 3 | 378 | |||
BX647566.1 | - | 8 | 1081 | |||
CR536528.1 | - | 7 | 983 | |||
CR541880.1 | - | 7 | 983 | |||
FJ474908.1 | - | 9 | 1150 | |||
M11560.1 | - | 9 | 1150 | |||
M21190.1 | - | 4 | 569 | |||
X05236.1 | - | 8 | 1129 |