Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029993.1 | NP_001025164.1 | 2 | 351 | ||
NM_001029994.1 | NP_001025165.1 | 2 | 351 | |||
GenBank mRNA | AK017933.1 | - | 2 | 328 | ||
AK031413.1 | - | 2 | 300 | |||
AK159881.1 | - | 2 | 287 | |||
AK166802.1 | - | 2 | 306 | |||
BC003319.1 | - | 1 | 199 | |||
BC030495.1 | - | 2 | 331 | 5400494 | ||
BY235998.1 | - | 2 | 358 | |||
CD348551.1 | - | 2 | 320 |