Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024274.3 | NP_077236.1 | 2 | 572 | ||
GenBank mRNA | AK013211.1 | - | 2 | 311 | ||
AK054370.1 | - | 337 | ||||
AK076259.1 | - | 2 | 356 | |||
AK170986.1 | - | 2 | 355 | |||
BC002147.1 | - | 2 | 342 | 3484842 |