Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024406.2 | NP_077717.1 | 2, 3 | 309 | ||
GenBank mRNA | AK003143.1 | - | 2, 3 | 310 | ||
AK088535.1 | - | 2, 3 | 310 | |||
BC054426.1 | - | 2, 3 | 292 | 5029359 | ||
BY081693.1 | - | 2, 3 | 309 | |||
CT010390.1 | - | 2, 3 | 244 | |||
K02109.1 | - | 2, 3 | 287 | |||
M28726.1 | - | 2, 3 | 214 |