Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024436.3 | NP_077756.2 | 5 | 618 | ||
GenBank mRNA | AB232616.1 | - | 5 | 357 | ||
AJ311124.1 | - | 5 | 414 | |||
AK014407.1 | - | 604 | ||||
AK031667.1 | - | 5 | 598 | |||
AK146594.1 | - | 5 | 618 | |||
AK147619.1 | - | 622 | ||||
AK149200.1 | - | 611 | ||||
AK150617.1 | - | 5 | 624 | |||
BC006596.1 | - | 5 | 605 | 3965264 |