Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Derl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024440.2 | NP_077760.1 | 4 | 317 | ||
GenBank mRNA | AB047555.1 | - | 4 | 317 | ||
AK007348.1 | - | 4 | 302 | |||
AK007948.1 | - | 4 | 303 | |||
AK172429.1 | - | 4 | 303 | |||
BC004729.1 | - | 4 | 300 | |||
BC019815.1 | - | 4 | 296 | |||
BC099615.1 | - | 4 | 297 | |||
BC115466.1 | - | 4 | 312 | 40100387 | ||
BC115467.1 | - | 4 | 312 | 40100388 |