Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024444.2 | NP_077764.2 | 3 | 270 | ||
GenBank mRNA | AF233647.1 | - | 3 | 261 | ||
AK007863.1 | - | 3 | 272 | |||
BC013494.1 | - | 3 | 257 | 4218398 |