Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cptp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024472.4 | NP_077792.2 | 2, 3 | 496 | ||
GenBank mRNA | AK040664.1 | - | 2, 3 | 497 | ||
AK214377.1 | - | 1, 2 | 18 | |||
BC002216.1 | - | 1, 2 | 188 | 3490501 |