Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039392.2 | NP_001034481.1 | 2 | 339 | ||
NM_001190327.1 | NP_001177256.1 | 4 | 350 | |||
NM_025284.4 | NP_079560.1 | 3 | 278 | |||
GenBank mRNA | AK003178.1 | - | 3 | 195 | ||
AK005512.1 | - | 3 | 195 | |||
AK008557.1 | - | 3 | 195 | |||
AK012361.1 | - | 3 | 195 | |||
AK019212.1 | - | 3 | 195 | |||
AK019232.1 | - | 3 | 196 | |||
AK131711.1 | - | 3 | 195 | |||
AK161987.1 | - | 3 | 195 | |||
BC037153.1 | - | 3 | 186 | |||
BC038926.1 | - | 3 | 196 | |||
BC048070.1 | - | 3 | 174 | |||
BC059777.1 | - | 3 | 182 | |||
BC070416.1 | - | 3 | 173 | |||
BC080312.1 | - | 3 | 176 | |||
BC089326.1 | - | 3 | 154 | |||
BC089327.1 | - | 2 | 134 | |||
BC092009.1 | - | 3 | 179 | |||
BC093587.1 | - | 3 | 180 | |||
BC094284.1 | - | 3 | 168 | |||
BC099374.1 | - | 3 | 177 | |||
BC145877.1 | - | 3 | 155 | 40131180 | ||
BC145879.1 | - | 3 | 155 | 40131182 | ||
CA466695.1 | - | 342 | ||||
CB273987.1 | - | 2 | 285 | |||
CJ042349.1 | - | 2 | 409 | |||
CJ042420.1 | - | 2 | 281 | |||
Z48496.1 | - | 2 | 272 |